siRNA Details
Gene Symbol: Ptrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178595.3 | NP_848710.1 | 5 | 855 | ||
GenBank mRNA | AK075833.1 | - | 5 | 856 | ||
BC098218.1 | - | 5 | 895 | 3466952 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178595.3 | NP_848710.1 | 5 | 855 | ||
GenBank mRNA | AK075833.1 | - | 5 | 856 | ||
BC098218.1 | - | 5 | 895 | 3466952 |