siRNA Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178599.4 | NP_848714.1 | 5 | 3308 | ||
| GenBank mRNA | AK077934.1 | - | 5 | 3308 | ||
| AK170011.1 | - | 5 | 3280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd8| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178599.4 | NP_848714.1 | 5 | 3308 | ||
| GenBank mRNA | AK077934.1 | - | 5 | 3308 | ||
| AK170011.1 | - | 5 | 3280 | |||