siRNA Details
| Gene Symbol: Rnf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178607.4 | NP_848722.1 | 6 | 2062 | ||
| GenBank mRNA | AK048813.1 | - | 6 | 2020 | ||
| AK089207.1 | - | 6 | 2049 | |||
| AK154093.1 | - | 6 | 2000 | |||
| AK165427.1 | - | 6 | 2020 | |||
| BC096529.1 | - | 6 | 1995 | 6406985 | ||

Rnf24
Genomic Map