siRNA Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077661.1 | NP_001071129.1 | 5 | 667 | ||
| NM_178623.3 | NP_848738.2 | 5 | 251 | |||
| GenBank mRNA | AK018583.1 | - | 1 | 58 | ||
| AK033577.1 | - | 5 | 225 | |||
| AK035580.1 | - | 5 | 663 | |||
| AK041953.1 | - | 6 | 916 | |||
| AK129378.1 | - | 4 | 309 | |||
| AK211902.1 | - | 1 | 18 | |||
| BC066994.1 | - | 5 | 548 | 5696000 | ||
| BY058620.1 | - | 5 | 251 | |||

Urgcp
Genomic Map