siRNA Details
| Gene Symbol: Tmem209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178625.3 | NP_848740.3 | 4, 5 | 354 | ||
| GenBank mRNA | AK029366.1 | - | 4, 5 | 364 | ||
| AK044877.1 | - | 4, 5 | 370 | |||
| AK078544.1 | - | 4, 5 | 368 | |||
| AK084222.1 | - | 3, 4 | 1908 | |||
| AK089486.1 | - | 229 | ||||
| AK089962.1 | - | 4, 5 | 369 | |||
| AK170465.1 | - | 229 | ||||
| BC052187.1 | - | 4, 5 | 397 | 6393580 | ||
| BC057109.1 | - | 4, 5 | 354 | 6400080 | ||

Tmem209
Genomic Map