siRNA Details
| Gene Symbol: C230052I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178643.5 | NP_848758.1 | 1, 2 | 553 | ||
| GenBank mRNA | AK031566.1 | - | 1, 2 | 594 | ||
| AK040248.1 | - | 1, 2 | 579 | |||
| AK082457.1 | - | 1, 2 | 554 | |||
| AK153918.1 | - | 1, 2 | 500 | |||
| AK166481.1 | - | 1, 2 | 600 | |||
| BC096687.1 | - | 1, 2 | 405 | 1433264 | ||

C230052I12Rik
Genomic Map