siRNA Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178651.3 | NP_848766.2 | 1, 2 | 248 | ||
| GenBank mRNA | AK028885.1 | - | 1, 2 | 215 | ||
| AK035541.1 | - | 1, 2 | 235 | |||
| AK038559.1 | - | 1, 2 | 213 | |||
| AK082853.1 | - | 1, 2 | 250 | |||
| AK143277.1 | - | 1, 2 | 249 | |||
| AY682914.1 | - | 1, 2 | 241 | |||
| BC141008.1 | - | 1, 2 | 214 | 9055620 | ||

Slc30a9
Genomic Map