siRNA Details
| Gene Symbol: Pde12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178668.3 | NP_848783.3 | 1 | 701 | ||
| GenBank mRNA | AK082947.1 | - | 1 | 524 | ||
| AK088843.1 | - | 1 | 514 | |||
| AK143567.1 | - | 1 | 524 | |||
| AK155088.1 | - | 535 | ||||
| AK161083.1 | - | 1 | 741 | |||
| AK167708.1 | - | 1 | 499 | |||
| BC064450.1 | - | 1 | 701 | 6417852 | ||

Pde12
Genomic Map