siRNA Details
| Gene Symbol: 8030462N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178670.3 | NP_848785.2 | 2 | 455 | ||
| GenBank mRNA | AK033209.1 | - | 2 | 442 | ||
| AK046785.1 | - | 2 | 434 | |||
| AK087799.1 | - | 2 | 379 | |||
| AK139552.1 | - | 2 | 352 | |||
| AK163840.1 | - | 456 | ||||
| BB649463.1 | - | 2 | 456 | |||
| BC120889.2 | - | 1 | 165 | 40060928 | ||

8030462N17Rik
Genomic Map