siRNA Details
| Gene Symbol: Ubxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285928.1 | NP_001272857.1 | 4 | 3172 | ||
| NM_001285929.1 | NP_001272858.1 | 3 | 3061 | |||
| NM_001285930.1 | NP_001272859.1 | 3 | 2925 | |||
| NM_178671.5 | NP_848786.1 | 2 | 2794 | |||
| GenBank mRNA | AK046949.1 | - | 4 | 3173 | ||
| AK077656.1 | - | 2 | 2797 | |||
| AK133185.1 | - | 2 | 2780 | |||
| AK161245.1 | - | 2 | 2781 | |||
| AK161296.1 | - | 2 | 2757 | |||

Ubxn10
Genomic Map