siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 3 | 270 | ||
| GenBank mRNA | AK038916.1 | - | 3 | 332 | ||
| AK044808.1 | - | 3 | 287 | |||
| AK044952.1 | - | 3 | 264 | |||
| AK047594.1 | - | 3 | 315 | |||
| AK079319.1 | - | 3 | 287 | |||
| AK089236.1 | - | 3 | 277 | |||
| AK089242.1 | - | 3 | 277 | |||
| BC053439.1 | - | 3 | 270 | 6328829 | ||

Cep120
Genomic Map