siRNA Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178704.3 | NP_848819.2 | 3 | 366 | ||
| GenBank mRNA | AF538322.1 | - | 3 | 350 | ||
| AK031345.1 | - | 3 | 358 | |||
| AK033395.1 | - | 3 | 323 | |||
| AK034217.1 | - | 3 | 367 | |||
| AK045758.1 | - | 3 | 267 | |||
| BC050065.1 | - | 3 | 310 | 30016211 | ||
| BC059897.1 | - | 3 | 263 | 6821433 | ||

Dpy19l3
Genomic Map