siRNA Details
| Gene Symbol: Cnpy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178612.4 | NP_848727.1 | 1 | 157 | ||
| GenBank mRNA | AK011474.1 | - | 1 | 158 | ||
| AK012968.1 | - | 1 | 139 | |||
| AK033219.1 | - | 1 | 159 | |||
| AK051562.1 | - | 1 | 159 | |||
| BC086647.1 | - | 1 | 122 | 6818372 | ||
| BC094672.1 | - | 1 | 264 | 30633103 | ||

Cnpy4
Genomic Map