siRNA Details
| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034856.1 | - | 2 | 252 | ||
| AK085362.1 | - | 2 | 289 | |||
| AK086462.1 | - | 2 | 282 | |||
| BC054120.1 | - | 2 | 235 | 6506658 | ||
| CD348032.1 | - | 2 | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints7| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034856.1 | - | 2 | 252 | ||
| AK085362.1 | - | 2 | 289 | |||
| AK086462.1 | - | 2 | 282 | |||
| BC054120.1 | - | 2 | 235 | 6506658 | ||
| CD348032.1 | - | 2 | 379 | |||