siRNA Details
| Gene Symbol: Bend7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190400.1 | NP_001177329.1 | 3 | 624 | ||
| NM_178663.4 | NP_848778.1 | 3 | 624 | |||
| GenBank mRNA | AK030428.1 | - | 3 | 311 | ||
| AK042822.1 | - | 3 | 622 | |||
| AK053896.1 | - | 3 | 626 | |||
| BC096430.1 | - | 3 | 296 | 5041388 | ||
| BC119370.1 | - | 3 | 263 | 8734119 | ||
| BC144864.1 | - | 3 | 263 | 9053412 | ||

Bend7
Genomic Map