siRNA Details
| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253719.1 | NP_001240648.1 | 2, 3 | 307 | ||
| NM_178673.4 | NP_848788.2 | 2, 3 | 567 | |||
| GenBank mRNA | AF374460.1 | - | 2, 3 | 541 | ||
| AK033652.1 | - | 2, 3 | 558 | |||
| AK046070.1 | - | 2, 3 | 308 | |||
| AK081152.1 | - | 2, 3 | 558 | |||
| AK138388.1 | - | 2, 3 | 859 | |||
| AK140611.1 | - | 2, 3 | 570 | |||
| BC075640.1 | - | 2, 3 | 530 | 30545529 | ||

Fstl5
Genomic Map