siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 4 | 352 | ||
| GenBank mRNA | AK038916.1 | - | 4 | 414 | ||
| AK044808.1 | - | 4 | 369 | |||
| AK044952.1 | - | 4 | 346 | |||
| AK047594.1 | - | 4 | 397 | |||
| AK079319.1 | - | 4 | 369 | |||
| AK089236.1 | - | 4 | 359 | |||
| AK089242.1 | - | 4 | 359 | |||
| BC053439.1 | - | 4 | 352 | 6328829 | ||

Cep120
Genomic Map