siRNA Details
| Gene Symbol: CXCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001511.3 | NP_001502.1 | 4 | 621 | ||
| NR_046035.1 | 4 | 644 | ||||
| GenBank mRNA | BC011976.1 | - | 4 | 609 | 3856841 | |
| BM554448.1 | - | 4 | 634 | |||
| J03561.1 | - | 4 | 595 | |||
| X12510.1 | - | 4 | 559 | |||

CXCL1
Genomic Map