siRNA Details
| Gene Symbol: Luzp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178705.5 | NP_848820.4 | 3, 4 | 973 | ||
| GenBank mRNA | AK032204.1 | - | 3, 4 | 630 | ||
| AK034077.1 | - | 3, 4 | 591 | |||
| AK034240.1 | - | 3, 4 | 625 | |||
| AK082126.1 | - | 3, 4 | 721 | |||
| AY164459.1 | - | 3, 4 | 970 | |||
| BC075646.1 | - | 3, 4 | 603 | 30620475 | ||

Luzp2
Genomic Map