siRNA Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178719.5 | NP_848834.2 | 5 | 835 | ||
| GenBank mRNA | AK038466.1 | - | 5 | 808 | ||
| AK044773.1 | - | 6 | 1268 | |||
| AK050546.1 | - | 5 | 796 | |||
| AK080375.1 | - | 5 | 781 | |||
| AK163389.1 | - | 6 | 936 | |||
| BC094597.1 | - | 5 | 767 | |||
| BC137633.1 | - | 4 | 739 | 8860652 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001007709, XM_006242081, XM_008765718, XM_006242083, XM_008765717, and others.

Mief1
Genomic Map