siRNA Details
| Gene Symbol: Cnpy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178612.4 | NP_848727.1 | 6 | 1126 | ||
| GenBank mRNA | AK012968.1 | - | 6 | 1108 | ||
| AK033219.1 | - | 6 | 1128 | |||
| AK051562.1 | - | 6 | 1128 | |||
| BC094672.1 | - | 6 | 1233 | 30633103 | ||

Cnpy4
Genomic Map