siRNA Details
Gene Symbol: Fam83g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178618.3 | NP_848733.2 | 4 | 1005 | ||
GenBank mRNA | AK028566.1 | - | 4 | 952 | ||
BC085279.1 | - | 4 | 966 | 30438273 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam83g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178618.3 | NP_848733.2 | 4 | 1005 | ||
GenBank mRNA | AK028566.1 | - | 4 | 952 | ||
BC085279.1 | - | 4 | 966 | 30438273 |