siRNA Details
| Gene Symbol: Tmem209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178625.3 | NP_848740.3 | 13, 14 | 1584 | ||
| GenBank mRNA | AK029366.1 | - | 10, 11 | 1273 | ||
| AK044877.1 | - | 13, 14 | 1600 | |||
| AK078544.1 | - | 13, 14 | 1598 | |||
| AK089486.1 | - | 1459 | ||||
| AK089962.1 | - | 13, 14 | 1599 | |||
| AK170465.1 | - | 1459 | ||||
| BC052187.1 | - | 13, 14 | 1627 | 6393580 | ||
| BC057109.1 | - | 13, 14 | 1584 | 6400080 | ||

Tmem209
Genomic Map