siRNA Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163328.1 | NP_001156800.1 | 9 | 1909 | ||
| NM_001163329.1 | NP_001156801.1 | 8 | 1502 | |||
| NM_001163330.1 | NP_001156802.1 | 7 | 1329 | |||
| NM_178631.4 | NP_848746.1 | 10 | 2002 | |||
| GenBank mRNA | AK032086.1 | - | 9 | 1842 | ||
| AK038465.1 | - | 8 | 1480 | |||
| AK079659.1 | - | 9 | 2731 | |||
| BC052358.1 | - | 8 | 1463 | 6534677 | ||

Ralyl
Genomic Map