siRNA Details
| Gene Symbol: Tigd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178646.4 | NP_848761.2 | 1 | 2915 | ||
| GenBank mRNA | AK036241.1 | - | 1 | 2890 | ||
| BC099838.1 | - | 1 | 2915 | 5703010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tigd5| Gene Symbol: Tigd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178646.4 | NP_848761.2 | 1 | 2915 | ||
| GenBank mRNA | AK036241.1 | - | 1 | 2890 | ||
| BC099838.1 | - | 1 | 2915 | 5703010 | ||