siRNA Details
| Gene Symbol: Oog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB077322.2 | - | 1 | 890 | ||
| AK145346.1 | - | 3 | 1280 | |||
| Gene Symbol: Gm2042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139641.1 | - | 3 | 1277 | ||

Oog1
Genomic Map