siRNA Details
| Gene Symbol: 8030462N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178670.3 | NP_848785.2 | 5 | 3051 | ||
| GenBank mRNA | AK087799.1 | - | 5 | 2981 | ||
| BC120889.2 | - | 4 | 2761 | 40060928 | ||
| DQ465967.1 | - | 1 | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8030462N17Rik| Gene Symbol: 8030462N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178670.3 | NP_848785.2 | 5 | 3051 | ||
| GenBank mRNA | AK087799.1 | - | 5 | 2981 | ||
| BC120889.2 | - | 4 | 2761 | 40060928 | ||
| DQ465967.1 | - | 1 | 696 | |||