siRNA Details
| Gene Symbol: Radil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289588.1 | NP_001276517.1 | 2 | 670 | ||
| NM_178702.4 | NP_848817.2 | 2 | 670 | |||
| GenBank mRNA | AK038570.1 | - | 2 | 603 | ||
| AK052964.1 | - | 2 | 581 | |||
| AK083410.1 | - | 2 | 688 | |||
| AK154169.1 | - | 2 | 610 | |||
| AK162917.1 | - | 2 | 650 | |||
| AK163852.1 | - | 2 | 606 | |||
| AK173277.1 | - | 2 | 634 | |||
| BC056483.1 | - | 2 | 622 | 5696312 | ||

Radil
Genomic Map