siRNA Details
| Gene Symbol: Tmem30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178715.3 | NP_848830.1 | 1 | 2698 | ||
| GenBank mRNA | AK033580.1 | - | 1 | 2699 | ||
| AK037246.1 | - | 1 | 2696 | |||
| AK169014.1 | - | 1 | 2734 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem30b| Gene Symbol: Tmem30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178715.3 | NP_848830.1 | 1 | 2698 | ||
| GenBank mRNA | AK033580.1 | - | 1 | 2699 | ||
| AK037246.1 | - | 1 | 2696 | |||
| AK169014.1 | - | 1 | 2734 | |||