siRNA Details
| Gene Symbol: ACTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104.3 | NP_001095.2 | 15 | 1799 | ||
| NM_001258371.2 | NP_001245300.2 | 15 | 1947 | |||
| NR_047663.2 | 15 | 1799 | ||||
| GenBank mRNA | AK125851.1 | - | 15 | 2333 | ||
| AK313556.1 | - | 15 | 1737 | |||
| BC096323.2 | - | 9 | 859 | |||
| BC099647.3 | - | 15 | 1720 | 40008220 | ||
| BC099648.1 | - | 5 | 468 | |||
| BC099649.3 | - | 15 | 1720 | 40008223 | ||
| DA827389.1 | - | 4 | 383 | |||
| M86407.1 | - | 15 | 1720 | |||

ACTN3
Genomic Map