siRNA Details
| Gene Symbol: ACVR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004302.4 | NP_004293.1 | 5, 6 | 1067 | ||
| NM_020327.3 | NP_064732.3 | 5, 6 | 1019 | |||
| NM_020328.3 | NP_064733.3 | 6, 7 | 1190 | |||
| XM_011538966.2 | XP_011537268.1 | 6, 7 | 1168 | |||
| XM_017020199.1 | XP_016875688.1 | 6, 7 | 1888 | |||
| XM_017020200.1 | XP_016875689.1 | 7, 8 | 1283 | |||
| XM_017020201.1 | XP_016875690.1 | 5, 6 | 1045 | |||
| GenBank mRNA | AK299120.1 | - | 6, 7 | 1153 | ||
| AK299496.1 | - | 5, 6 | 1019 | |||
| BC000254.1 | - | 5, 6 | 1016 | 3352720 | ||
| BC040531.1 | - | 5, 6 | 1064 | 5260767 | ||
| BT007072.1 | - | 5, 6 | 990 | |||
| L10125.1 | - | 5, 6 | 1029 | |||
| L10126.1 | - | 5, 6 | 990 | |||
| U14722.1 | - | 5, 6 | 990 | |||
| Z22536.1 | - | 5, 6 | 990 | |||

ACVR1B
Genomic Map