siRNA Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106.3 | NP_001097.2 | 7, 8 | 977 | ||
| XM_005265583.3 | XP_005265640.1 | 7, 8 | 3439 | |||
| XM_017007514.1 | XP_016863003.1 | 7, 8 | 3418 | |||
| XM_017007515.1 | XP_016863004.1 | 7, 8 | 1063 | |||
| XM_017007516.1 | XP_016863005.1 | 7, 8 | 4387 | |||
| GenBank mRNA | AB209449.1 | - | 6, 7 | 4742 | ||
| BC096243.1 | - | 7, 8 | 957 | 40005760 | ||
| BC096244.1 | - | 7, 8 | 957 | 40005761 | ||
| BC096245.1 | - | 6, 7 | 1039 | 40005763 | ||
| BC099642.4 | - | 7, 8 | 957 | 40005762 | ||
| X77533.1 | - | 7, 8 | 957 | |||

ACVR2B
Genomic Map