siRNA Details
| Gene Symbol: ADH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000667.3 | NP_000658.1 | 3, 4 | 383 | ||
| GenBank mRNA | AK290558.1 | - | 3, 4 | 340 | ||
| AK309597.1 | - | 3, 4 | 358 | |||
| BC074738.2 | - | 3, 4 | 297 | 30915195 | ||
| BC117442.1 | - | 3, 4 | 339 | 40125993 | ||
| BC126306.1 | - | 3, 4 | 339 | 8992022 | ||
| BT019812.1 | - | 3, 4 | 269 | |||
| M12271.1 | - | 3, 4 | 284 | |||
| M12963.1 | - | 3, 4 | 341 | |||

ADH1A
Genomic Map