siRNA Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000672.3 | NP_000663.1 | 2 | 146 | ||
| NM_001102470.1 | NP_001095940.1 | 2 | 146 | |||
| NR_132990.1 | 2 | 146 | ||||
| GenBank mRNA | AK092768.1 | - | 2 | 146 | ||
| AK316326.1 | - | 2 | 74 | |||
| BC039065.1 | - | 2 | 135 | 5184104 | ||

ADH6
Genomic Map