siRNA Details
| Gene Symbol: AEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129.4 | NP_001120.3 | 17, 18 | 2512 | ||
| XM_011515162.1 | XP_011513464.1 | 16, 17 | 2475 | |||
| GenBank mRNA | AB209744.1 | - | 17, 18 | 2466 | ||
| AF053944.1 | - | 17, 18 | 2346 | |||
| AK090931.1 | - | 4, 5 | 604 | |||
| AK127541.1 | - | 4, 5 | 1000 | |||
| AK296001.1 | - | 3, 4 | 506 | |||
| BC038588.1 | - | 17, 18 | 2512 | 5764769 | ||
| D86479.1 | - | 11, 12 | 1268 | |||

AEBP1
Genomic Map