siRNA Details
| Gene Symbol: AEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129.4 | NP_001120.3 | 18 | 2579 | ||
| XM_011515162.1 | XP_011513464.1 | 17 | 2542 | |||
| GenBank mRNA | AB209744.1 | - | 18 | 2533 | ||
| AF053944.1 | - | 18 | 2413 | |||
| AK127541.1 | - | 5 | 1067 | |||
| AK296001.1 | - | 4 | 573 | |||
| BC038588.1 | - | 18 | 2579 | 5764769 | ||
| D86479.1 | - | 12 | 1335 | |||

AEBP1
Genomic Map