siRNA Details
| Gene Symbol: CRISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131.2 | NP_001122.2 | 4, 5 | 355 | ||
| NM_001205220.1 | NP_001192149.1 | 4, 5 | 366 | |||
| NM_170609.1 | NP_733758.1 | 4, 5 | 355 | |||
| GenBank mRNA | AB451334.1 | - | 3, 4 | 276 | ||
| AB451479.1 | - | 3, 4 | 276 | |||
| AK292497.1 | - | 4, 5 | 336 | |||
| BC131707.1 | - | 4, 5 | 326 | |||
| GQ472203.1 | - | 4, 5 | 349 | |||
| X95237.1 | - | 4, 5 | 349 | |||
| X95238.1 | - | 4, 5 | 349 | |||

CRISP1
Genomic Map