siRNA Details
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 6 | 712 | |||
NR_003227.1 | 6 | 639 | ||||
NR_003228.1 | 6 | 678 | ||||
GenBank mRNA | AF329691.1 | - | 5 | 463 | ||
AF329693.1 | - | 5 | 463 | |||
AF329694.1 | - | 4 | 394 | |||
AK056488.1 | - | 6 | 665 | |||
AK091293.1 | - | 6 | 675 | |||
AK097044.1 | - | 6 | 626 | |||
AK126911.1 | - | 5 | 603 | |||
AK128804.1 | - | 3 | 251 | |||
BC021245.2 | - | 4 | 285 |
