siRNA Details
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 9 | 1094 | |||
NR_003227.1 | 8 | 921 | ||||
NR_003228.1 | 9 | 1060 | ||||
GenBank mRNA | AF329691.1 | - | 8 | 845 | ||
AF329692.1 | - | 3 | 396 | |||
AJ001495.1 | - | 1 | 68 | |||
AK056488.1 | - | 9 | 1065 | |||
AK091293.1 | - | 9 | 1057 | |||
AK097044.1 | - | 8 | 908 | |||
AK126911.1 | - | 7 | 885 | |||
AK128352.1 | - | 2 | 760 | |||
AL832984.1 | - | 3 | 318 | |||
BC021245.2 | - | 7 | 667 |
