siRNA Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000027.3 | NP_000018.2 | 7 | 911 | ||
| NM_001171988.1 | NP_001165459.1 | 7 | 881 | |||
| NR_033655.1 | 6 | 835 | ||||
| GenBank mRNA | AK312982.1 | - | 7 | 845 | ||
| AW467768.1 | - | 2 | 154 | |||
| BC012392.1 | - | 7 | 834 | 3859317 | ||
| CR541715.1 | - | 7 | 783 | |||
| CX869892.1 | - | 6 | 712 | |||
| M64073.1 | - | 7 | 786 | |||
| X55762.1 | - | 7 | 810 | |||

AGA
Genomic Map