siRNA Details
| Gene Symbol: ALDH1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000689.4 | NP_000680.2 | 11, 12 | 1673 | ||
| GenBank mRNA | AB209821.1 | - | 11, 12 | 1400 | ||
| AF003341.1 | - | 11, 12 | 1356 | |||
| AK000118.1 | - | 11, 12 | 1400 | |||
| AK026641.1 | - | 11, 12 | 1400 | |||
| AK293176.1 | - | 11, 12 | 1366 | |||
| AK302168.1 | - | 10, 11 | 1184 | |||
| AY390731.1 | - | 11, 12 | 1356 | |||
| BC001505.2 | - | 11, 12 | 1381 | 2988388 | ||
| BT006921.1 | - | 11, 12 | 1356 | |||
| EU668318.1 | - | 11, 12 | 1400 | |||
| EU794589.1 | - | 11, 12 | 1356 | |||
| FJ224286.1 | - | 11, 12 | 1409 | |||
| K03000.1 | - | 7, 8 | 873 | |||
| M26761.1 | - | 7, 8 | 873 | |||

ALDH1A1
Genomic Map