siRNA Details
| Gene Symbol: ALPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001631.4 | NP_001622.2 | 9 | 1099 | ||
| GenBank mRNA | BC132678.1 | - | 9 | 1041 | 40146700 | |
| M15694.1 | - | 9 | 1011 | |||
| M31008.1 | - | 8 | 1101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALPI| Gene Symbol: ALPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001631.4 | NP_001622.2 | 9 | 1099 | ||
| GenBank mRNA | BC132678.1 | - | 9 | 1041 | 40146700 | |
| M15694.1 | - | 9 | 1011 | |||
| M31008.1 | - | 8 | 1101 | |||