siRNA Details
| Gene Symbol: AMBN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016519.5 | NP_057603.1 | 12 | 871 | ||
| GenBank mRNA | AF209780.1 | - | 12 | 853 | ||
| AF219994.2 | - | 12 | 835 | |||
| AF263464.1 | - | 12 | 845 | |||
| AJ537437.1 | - | 12 | 845 | |||
| AK314918.1 | - | 12 | 806 | |||
| BC106931.1 | - | 12 | 806 | 40033703 | ||

AMBN
Genomic Map