siRNA Details
| Gene Symbol: AMBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001633.3 | NP_001624.1 | 3 | 587 | ||
| GenBank mRNA | AK290837.1 | - | 3 | 415 | ||
| AK303795.1 | - | 3 | 313 | |||
| AK310776.1 | - | 3 | 514 | |||
| AK310791.1 | - | 3 | 443 | |||
| AY544123.1 | - | 3 | 384 | |||
| BC041593.1 | - | 3 | 550 | 5433460 | ||
| X04225.1 | - | 3 | 396 | |||
| X04494.1 | - | 3 | 369 | |||

AMBP
Genomic Map