siRNA Details
| Gene Symbol: APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000483.4 | NP_000474.2 | 3, 4 | 313 | ||
| GenBank mRNA | AA883870.1 | - | 1, 2 | 422 | ||
| BC005348.2 | - | 2, 3 | 219 | 3935036 | ||
| BT006708.1 | - | 2, 3 | 206 | |||
| BU596546.1 | - | 3, 4 | 349 | |||
| KU177908.1 | - | 2, 3 | 206 | |||
| KU177909.1 | - | 2, 3 | 242 | |||
| M29844.1 | - | 2, 3 | 176 | |||
| X00568.1 | - | 2, 3 | 206 | |||
| Gene Symbol: APOC4-APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037932.1 | 5, 6 | 1413 | |||
| GenBank mRNA | AK126257.1 | - | 6, 7 | 1347 | ||

APOC2
Genomic Map