siRNA Details
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042681.1 | NP_001036146.1 | 13, 14 | 2067 | ||
| NM_001042682.1 | NP_001036147.1 | 3, 4 | 366 | |||
| NM_012102.3 | NP_036234.3 | 14, 15 | 2252 | |||
| XM_005263464.2 | XP_005263521.1 | 13, 14 | 2006 | |||
| XM_005263466.1 | XP_005263523.1 | 7, 8 | 889 | |||
| XM_011541510.1 | XP_011539812.1 | 12, 13 | 1992 | |||
| XM_011541511.1 | XP_011539813.1 | 13, 14 | 2118 | |||
| XM_017001358.1 | XP_016856847.1 | 13, 14 | 1757 | |||
| XM_017001359.1 | XP_016856848.1 | 13, 14 | 2262 | |||
| GenBank mRNA | AB007927.2 | - | 12, 13 | 1402 | ||
| AB036737.1 | - | 14, 15 | 2078 | |||
| AF016005.1 | - | 3, 4 | 345 | |||

RERE
Genomic Map