siRNA Details
| Gene Symbol: ATP2B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001344.2 | NP_001001344.1 | 16 | 2793 | ||
| NM_021949.3 | NP_068768.2 | 16 | 2793 | |||
| XM_005274689.3 | XP_005274746.1 | 18 | 3111 | |||
| XM_005274690.3 | XP_005274747.1 | 18 | 3111 | |||
| XM_005274691.3 | XP_005274748.1 | 17 | 3069 | |||
| XM_005274692.3 | XP_005274749.1 | 18 | 3101 | |||
| XM_011531174.2 | XP_011529476.1 | 18 | 3112 | |||
| XM_011531177.2 | XP_011529479.1 | 17 | 3070 | |||
| XM_017029553.1 | XP_016885042.1 | 17 | 3015 | |||
| GenBank mRNA | BC130009.1 | - | 16 | 2726 | 40067815 | |
| BQ878721.1 | - | 151 | ||||
| BU183973.1 | - | 4 | 580 | |||
| U57971.1 | - | 16 | 2793 | |||
| U60414.1 | - | 16 | 2793 | |||

ATP2B3
Genomic Map