siRNA Details
| Gene Symbol: ADGRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001704.2 | NP_001695.1 | 22 | 3444 | ||
| XM_005248752.2 | XP_005248809.1 | 21 | 3730 | |||
| XM_006715534.3 | XP_006715597.1 | 22 | 3866 | |||
| XM_011536009.2 | XP_011534311.1 | 20 | 2329 | |||
| XM_011536010.2 | XP_011534312.1 | 22 | 3503 | |||
| XM_011536011.2 | XP_011534313.1 | 21 | 3734 | |||
| XM_017011140.1 | XP_016866629.1 | 21 | 3694 | |||
| XM_017011141.1 | XP_016866630.1 | 19 | 2201 | |||
| GenBank mRNA | AB005299.1 | - | 21 | 3029 | ||
| AB011122.2 | - | 22 | 3233 | |||
| AK289696.1 | - | 21 | 3045 | |||
| AK295516.1 | - | 6 | 740 | |||
| AK316174.1 | - | 6 | 652 | |||
| BC111720.1 | - | 20 | 3023 | 40027602 | ||
More Information
Set Membership: |
|

ADGRB3
Genomic Map