siRNA Details
| Gene Symbol: NKX3-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001189.3 | NP_001180.1 | 2 | 1011 | ||
| GenBank mRNA | AF009801.1 | - | 2 | 935 | ||
| BC111966.1 | - | 2 | 1011 | 8327434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX3-2| Gene Symbol: NKX3-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001189.3 | NP_001180.1 | 2 | 1011 | ||
| GenBank mRNA | AF009801.1 | - | 2 | 935 | ||
| BC111966.1 | - | 2 | 1011 | 8327434 | ||