siRNA Details
| Gene Symbol: BBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031885.3 | NP_114091.3 | 8 | 1074 | ||
| XM_005256080.2 | XP_005256137.1 | 8 | 1073 | |||
| GenBank mRNA | AB208905.1 | - | 8 | 854 | ||
| AF342736.1 | - | 8 | 1261 | |||
| AK027635.1 | - | 8 | 1002 | |||
| AK289604.1 | - | 8 | 964 | |||
| AK309068.1 | - | 8 | 974 | |||
| BC014140.2 | - | 8 | 964 | 4653315 | ||

BBS2
Genomic Map